D(+)-Biotin

Lieferant: Biotium

Synonyme: (3aS,4S,6aR)-Biotin, Vitamin H, Vitamin B8, 5-[(3aS,4S,6aR)-2-Oxohexahydro-1H- thieno[3,4-d]imidazol-4-yl]pentansäure, Vitamin B7, Coenzym R

90072
90072.EA 79.6 EUR
90072.
D(+)-Biotin
D(+)-Biotin
Formel: C₁₀H₁₆N₂O₃S
Molecular Weight: 244,31 g/mol
Schmelzpunkt: 232…233 °C
Dichte: 1,41 g/cm³ (20 °C)
Lagertemperatur: Kühlschrank
MDL: MFCD00005541
CAS-Nummer: 58-85-5
EINECS: 200-399-3
Merck Index: 13,01231

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR