Biotin ethylenediamine hydrochloride

Lieferant: Biotium

Synonyme: (3aS,4S,6aR)-N-(2-Aminoethyl)hexahydro- 2-oxo-1H-thieno[3,4-d]imidazole-4-pentanamide hydrochloride, N-(2-Aminoethyl)biotinamide hydrochloride

90075
90075.EA 146 EUR
90075.
Biotin ethylenediamine hydrochloride
Biotin ethylenediamine hydrochloride

Biotin ethylenediamine, hydrochloride is functionally equivalent to Neurobiotin™ (biotin ethylenediamine, hydrobromide, 90057) , which is a useful anterograde and transneuronal tracers.


Neurobiotin is a trademark of Vector Laboratories.

Formel: C₁₂H₂₃ClN₄O₂S
Molecular Weight: 322,5 g/mol
Lagertemperatur: Kühlschrank
CAS-Nummer: 216299-38-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a066452b1f9455a26c7d385f85c13432dd1ac622&vid=df6d61bb87a3719a16174fb51f400735d8c606bf&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR