Aceton ≥99,5% NF, FCC, J.T.Baker®

Lieferant: Avantor
Gefahr

Synonyme: Propanon, DMK, 2-Propanon, Dimethylketon

9008-01EA 45.8 EUR
9008-01 9008-03 9008-R
Aceton ≥99,5% NF, FCC, J.T.Baker®
Aceton
Formel: CH₃COCH₃
Siedepunkt: 56,2 °C (1013 hPa)
Schmelzpunkt: –95,4 °C
Dichte: 0,792 g/cm³ (20 °C)
Flammpunkt: <–20 °C (geschlossener Tiegel)
Lagertemperatur: Raumtemperatur
MDL: MFCD00008765
CAS-Nummer: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3,II
REACH: 01-2119471330-49
Merck Index: 13,00067
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Food GMP Manufactured Product
GMP Manufactured Product
Meets F.C.C. Requirements
Meets N.F. Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Food Chemical
Bulk Pharmaceutical Chemical Passes Test
NF - Identification A Passes Test
NF - Identification B Passes Test
NF - Benzene ≤ 2 ppm
NF - Specific Gravity at 25°/25°C ≤ 0.788
NF - Nonvolatile Residue ≤ 10 ppm
NF - Readily Oxidizable Substances Passes Test
NF - Water (H₂O) ≤ 0.5 %
NF - Assay (as (CH₃)₂CO) (calculated on anhydrous basis) 99.5 - 100.5 %
FCC - Identification A Passes Test
FCC - Identification B Passes Test
FCC - Solubility in H₂O Passes Test
FCC - Substances Reducing Permanganate Passes Test
FCC - Acidity (as CH₃COOH) ≤ 0.002 %
FCC - Aldehydes (as HCHO) ≤ 0.002 %
FCC - Alkalinity (as NH₃) ≤ 10 ppm
FCC - Lead (Pb) ≤ 1 mg/kg
FCC - Methanol (CH₃OH) ≤ 0.05 %
FCC - Phenols Passes Test
FCC - Refractive Index at 20°C 1.358 - 1.360
FCC - Nonvolatile Residue ≤ 10 ppm
FCC - Water (H₂O) ≤ 0.5 %
FCC - Distilling Range ≤ 1 °C
FCC - Initial Boiling Point
FCC - Dry Point
Class 1, 2 and 3 residual solvents are likely to be present. Benzene (Class 1) is reported on each lot and is below the USP/NF limit concentration. Methanol and Toluene (Class 2) are both below their
Option 1 limits. All Class 3 solvents are less than 0.5%.
Elemental Impurities (USP (232), EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR