Dimethylsulfoxid, wasserfrei ≥99,9%
Lieferant: Biotium
Anhydrous DMSO (dimethyl sulfoxide; methyl sulfoxide) is recommended for preparing stock solutions.
- Prepare stock solutions of AM ester dyes
- Stock solutions of reactive dyes and other related, moisture-sensitive products
Anhydrous DMSO (dimethyl sulfoxide; methyl sulfoxide) is recommended for preparing stock solutions of AM ester dyes, reactive dyes and other related moisture-sensitive products.
Formel:
(CH₃)₂SO Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002089 CAS-Nummer: 67-68-5 |
|
Spezifikation Testergebnisse
Color & Form | Colorless liquid |
Purity | ≥ 99.9% |
Water | Max 0.005% |
Residual on evaporation | Max 0.0005% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...