5-FAM-PEO₈ SE Amin-reaktiver fluoreszierender Farbstoff

Lieferant: Biotium

Synonyme: FAM-PEO8 SE, 5 isomer, 5-carboxyfluorescein-PEO8 propionate succinimidyl ester, 5-FAM-PEO8 SE

90085
90085.EA 232 EUR
90085.
5-FAM-PEO₈ SE Amin-reaktiver fluoreszierender Farbstoff
5-FAM-PEO₈ SE

5-FAM-PEO8 SE (full name: 5-Carboxyfluorescein-PEO8 propionate succinimidyl ester) has an approximately 32 Angstrom water-soluble PEG spacer that separates fluorescein and the amine-reactive SE group.


  • λEx/λ Em(pH9) = 494/521 nm
  • Yellow orange solid soluble in DMF, DMSO, or pH 6.5 buffer
  • Store at -20°C and protect from light

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5f0a79c2ad634a82203a23deedeb940e39603f00&vid=0cbe7be884347e422365a0d70a1bb7d79d42da4c&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR