5-Carboxy-X-rhodamine-PEO₈-propionate succinimidyl ester Amin-reaktiver fluoreszierender Farbstoff

Lieferant: Biotium

Synonyme: 5-ROX-PEO8, 5-Carboxy-X-rhodamine-PEO8-propionate succinimidyl ester, ROX-PEO8 SE, 5 isomer

90089
90089.EA 232 EUR
90089.
5-Carboxy-X-rhodamine-PEO₈-propionate succinimidyl ester Amin-reaktiver fluoreszierender Farbstoff
5-Carboxy-X-rhodamine-PEO₈-propionate succinimidyl ester

5-ROX-PEO8 SE (full name: 5-Carboxy-X-rhodamine-PEO8-propionate, succinimidyl ester) has an approximately 32 Angstrom water-soluble PEG spacer that separates the red fluorescent ROX dye and the amine-reactive SE group.


  • λEx/λ Em(MeOH) = 568/595 nm
  • Dark red solid soluble in DMSO or DMF
  • Store at -20°C and protect from light

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6102fa9126483aff5639bf0addf6e78bf293ab17&vid=f593345109d974d4db491d78140a470fca80d80d&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR