Aceton ≥99,5%, BAKER ANALYZED®, elektronische Qualität, J.T.Baker®

Lieferant: Avantor
Gefahr

Synonyme: Propanon, DMK, 2-Propanon, Dimethylketon

9009-07EA 418 EUR
9009-07
Aceton ≥99,5%, BAKER ANALYZED®, elektronische Qualität, J.T.Baker®
Aceton
Formel: CH₃COCH₃
Molecular Weight: 58,08 g/mol
Siedepunkt: 56,2 °C (1013 hPa)
Schmelzpunkt: –95,4 °C
Dichte: 0,792 g/cm³ (20 °C)
Flammpunkt: <–20 °C (geschlossener Tiegel)
Lagertemperatur: Raumtemperatur
MDL: MFCD00008765
CAS-Nummer: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3,II
REACH: 01-2119471330-49
Merck Index: 13,00067
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

For Microelectronic Use = 99.5 %
Assay ((CH₃)₂CO) (by GC, corrected for water) ≥ 99.5 %
Color (APHA) ≤ 10
Residue after Evaporation ≤ 0.001 %
Solubility in H₂O Passes Test
Titrable Acid (meq/g) ≤ 0.0003
Titrable Base (meq/g) ≤ 0.0006
Aldehydes (as HCHO) ≤ 0.002 %
Isopropyl Alcohol (CH₃CHOHCH₃)(by GC) ≤ 0.05 %
Methanol (CH₃OH)(by GC) ≤ 0.05 %
Heavy Metals (as Pb) ≤ 0.20 ppm
Substances Reducing Permanganate Passes Test
Water (H₂O) ≤ 0.5 %
Chloride (Cl) ≤ 0.2 ppm
Copper (Cu) ≤ 0.1 ppm
Iron (Fe) ≤ 0.050 ppm
Nickel (Ni) ≤ 0.1 ppm
Manganese (Mn) ≤ 0.100 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR