Fluorescamin ≥95% (durch TLC) Amin-reaktiver fluoreszierender Farbstoff

Lieferant: Biotium

Synonyme: Fluram, 4-Phenyl-3H,3'H-spiro[furan-2,1'-isobenzofuran]-3,3'-dione, 4-Phenylspiro[furan-2(3H),1'-phthalan]-3,3'-dione

90092
90092.EA 206 EUR
90092.
Fluorescamin ≥95% (durch TLC) Amin-reaktiver fluoreszierender Farbstoff
Fluorescamine

Fluorescamine is nonfluorescent until it reacts with primary amines. The reaction is completed in miniseconds and can be used to detect amine-containing compounds including peptides and proteins. Unreacted fluorescamine is rapidly converted to a non-fluorescent product by reaction with water, making amine detection highly specific.


  • λEx/λ Em(CH3CN) = 381/470 nm (for reaction product with amines)
  • Off-white solid soluble in DMF or DMSO
  • Store at -20°C and protect from light

Formel: C₁₇H₁₀O₄
Molecular Weight: 278,26 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00005928
CAS-Nummer: 38183-12-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR