6-(Fluorescein-5-carboxamido) hexanoic acid succinimidyl ester (5-SFX) ≥95% (durch TLC) Amin-reaktiver fluoreszierender Farbstoff
Lieferant: Biotium
90095
90095.EA
210
EUR
90095.
6-(Fluorescein-5-carboxamido) hexanoic acid succinimidyl ester (5-SFX) ≥95% (durch TLC) Amin-reaktiver fluoreszierender Farbstoff
6-(Fluorescein-5-carboxamido) hexanoic acid succinimidyl ester (5-SFX)
5-SFX (full name: 6-(Fluorescein-5-carboxamido)hexanoic acid, succinimidyl ester, single isomer) has a 6-atom aminohexanoyl spacer group that separates the fluorescein dye and the amine-reactive group.
- λEx/λ Em(pH9) = 494/521nm
- Yellow solid soluble in DMF, DMSO or pH>6.5 buffers
- Store at -20°C and protect from light