6-(Fluorescein-5-carboxamido) hexanoic acid succinimidyl ester (5-SFX) ≥95% (durch TLC) Amin-reaktiver fluoreszierender Farbstoff
Lieferant: Biotium
90095
90095.EA 210 EUR
90095.
6-(Fluorescein-5-carboxamido) hexanoic acid succinimidyl ester (5-SFX) ≥95% (durch TLC) Amin-reaktiver fluoreszierender Farbstoff
6-(Fluorescein-5-carboxamido) hexanoic acid succinimidyl ester (5-SFX)
5-SFX (full name: 6-(Fluorescein-5-carboxamido)hexanoic acid, succinimidyl ester, single isomer) has a 6-atom aminohexanoyl spacer group that separates the fluorescein dye and the amine-reactive group.
  • λEx/λ Em(pH9) = 494/521nm
  • Yellow solid soluble in DMF, DMSO or pH>6.5 buffers
  • Store at -20°C and protect from light
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=054b65521ba3e31ceb12cb3b699a6d72d2b580e8&vid=054b65521ba3e31ceb12cb3b699a6d72d2b580e8&prid=web&clid=SAPDAM