6-(Fluorescein-6-carboxamido) hexanoic acid succinimidyl ester (6-SFX) Amin-reaktiver fluoreszierender Farbstoff
Lieferant: Biotium
90096
90096.EA 210 EUR
90096.
6-(Fluorescein-6-carboxamido) hexanoic acid succinimidyl ester (6-SFX) Amin-reaktiver fluoreszierender Farbstoff
6-(Fluorescein-6-carboxamido) hexanoic acid succinimidyl ester (6-SFX)
6-SFX (full name: 6-(Fluorescein-6-carboxamido)hexanoic acid, succinimidyl ester, single isomer) has a 6-atom aminohexanoyl spacer group that separates the fluorescein dye and the amine-reactive group.
  • λEx/λEm (pH 9) = 494/521 nm
  • Yellow solid soluble in DMF, DMSO or pH>6.5 buffers
  • Store at -20°C and protect from light
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fdcedbe090c4e568106d4eb837471c87ec25b49b&vid=fdcedbe090c4e568106d4eb837471c87ec25b49b&prid=web&clid=SAPDAM