6-(Fluorescein-6-carboxamido) hexanoic acid succinimidyl ester (6-SFX) Amin-reaktiver fluoreszierender Farbstoff
Lieferant: Biotium
90096
90096.EA
210
EUR
90096.
6-(Fluorescein-6-carboxamido) hexanoic acid succinimidyl ester (6-SFX) Amin-reaktiver fluoreszierender Farbstoff
6-(Fluorescein-6-carboxamido) hexanoic acid succinimidyl ester (6-SFX)
6-SFX (full name: 6-(Fluorescein-6-carboxamido)hexanoic acid, succinimidyl ester, single isomer) has a 6-atom aminohexanoyl spacer group that separates the fluorescein dye and the amine-reactive group.
- λEx/λEm (pH 9) = 494/521 nm
- Yellow solid soluble in DMF, DMSO or pH>6.5 buffers
- Store at -20°C and protect from light