6-Carboxytetramethylrhodamine succinimidyl ester (6-TAMRA SE) 90 mg/mL in anhydrischem DMSO Amin-reaktiver fluoreszierender Farbstoff

Lieferant: Biotium

Synonyme: 6-Carboxytetramethylrhodamine N-succinimidyl ester, 6-TAMRA, SE, 6-TAMRA N-succinimidyl ester

90097
90097.EA 463 EUR
90097.
6-Carboxytetramethylrhodamine succinimidyl ester (6-TAMRA SE) 90 mg/mL in anhydrischem DMSO Amin-reaktiver fluoreszierender Farbstoff
6-Carboxytetramethylrhodamine succinimidyl ester (6-TAMRA SE)

6-TAMRA SE (full name: 6-Carboxytetramethylrhodamine succinimidyl ester) is the amine-reactive form of Amine-reactive form of Carboxytetramethylrhodamine (TAMRA) single isomer. Supplied at 90 mg/mL (0.17 M) in anhydrous DMSO. The solution is suitable for labeling oligonucleotides.


  • λEx/λEm (MeOH) = 540/565 nm
  • Dark red solution
  • Store desiccated at -20°C and protect from light


The solution is good for at least one month if stored at -20°C and keep away from moisture.

Formel: C₂₉H₂₅N₃O₇
Molecular Weight: 527.52 g/mol
Lagertemperatur: Gefrierschrank
CAS-Nummer: 150810-69-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e9b6d0519573726def6c916165d9d07e8b74df7d&vid=e9b6d0519573726def6c916165d9d07e8b74df7d&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR