Sulphorhodamine 101-PEO₈-propionic acid succinimydyl ester Amin-reaktiver fluoreszierender Farbstoff
Lieferant: Biotium
Synonyme:
Texas Red-PEO8 SE, Sulphorhodamine 101-PEO8 -propionic acid succinimydyl ester, SR101-PEO8 SE
90099
90099.EA
232
EUR
90099.
Sulphorhodamine 101-PEO₈-propionic acid succinimydyl ester Amin-reaktiver fluoreszierender Farbstoff
Sulphorhodamine 101-PEO₈-propionic acid succinimydyl ester
SR101-PEO8 SE (Sulforhodamine 101-PEO8-propionic acid succinimydyl ester; Texas Red®-PEO8 SE) has an approximately 32 Angstrom water-soluble PEG spacer that separates the dye from the amine-reactive SE group.
- λEx/λ Em= 583/603nm
- Dark purple solid soluble in DMF or DMSO
- Store at -20°C and protect from light, especially in solution
Texas Red is a registered trademark of Thermo Fisher Scientific.
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...