Gram Jodlösung für die Mikroskopie, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY


M-Clarity™ Qualitätsstufe = MQ100

90107-2.5L-F 90107-250ML-F
90107-2.5L-FEA 91.6 EUR
90107-2.5L-F 90107-250ML-F
Gram Jodlösung für die Mikroskopie, Sigma-Aldrich®
Gram Jodlösung

Gram’s iodine solution is used as a mordant in the Gram staining process. It is added after the primary staining step with crystal violet; the resultant crystal violet - Iodine complex imparts purple color to the cells.


Iodine(I) interacts with Crystal Violet(CV) to form large CV-I complexes within the cytoplasm and outer layers of the cell.


Syn: Potassium Triiodide

Formel: I₃K
Dichte: 1,005 g/ml (20 °C)
Lagertemperatur: Room Temperature
MDL: MFCD00134128
CAS-Nummer: 12298-68-9

Bestellen

Spezifikation Testergebnisse

Appearance (colour) Brown to very dark brown
Appearance (form) Liquid
Density D20/4 1.003 - 1.007
Refractive index N20/D 1.334 - 1.336
Expiration date period 24 months


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR