Cyanine 555NS succinimidyl ester Cyaninfarbstoff, nicht-sulfoniert
Lieferant: Biotium
90118-1MG
90118-1MGEA 204 EUR
90118-1MG
Cyanine 555NS succinimidyl ester Cyaninfarbstoff, nicht-sulfoniert
Cyanine succinimidyl ester
Cyanine NS succinimidyl esters are non-sulfonated cyanine dyes that can be used to label proteins, oligonucleotides and nucleic acids. Available in 3 colors, Cyanine 488NS, Cyanine 555NS, and Cyanine 647NS are designed to match by charge and molecular weight for 2D DIGE platform.
  • Ex/Em 547/572 nm
  • MW added to protein: 441
  • Red solid soluble in DMF or DMSO
  • Store desiccated at -20°C; protect from light
Cyanine NS succinimidyl esters are non-sulfonated cyanine dyes that can be used to label proteins, oligonucleotides and nucleic acids. Available in 3 colors, Cyanine 488NS, Cyanine 555NS and Cyanine 647NS are non-sulfonated versions of Cy®2, Cy®3, and Cy®5, respectively. They are designed to match by charge and molecular weight for 2D DIGE platform. Biotium offers these dyes as high quality amine-reactive succinimidyl esters.
Also see sulfonated Cyanine Dye Succinimidyl Esters and our CF® Dye Succinimidyl Esters, available with more than 20 bright and photostable CF® Dyes.
Cy2, Cy3, Cy5 are registered trademarks of GE Healthcare.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b5212f02ea41a4118dc06487a1d345ec9283334a&vid=0a13d14e6ccd39e1636b32d40ff93a38c4b66042&prid=web&clid=SAPDAM