Acetonitril ≥99,9%, BAKER ANALYZED® HPLC, Ultragradient für die HPLC, UHPLC, für die Spektrophotometrie, J.T.Baker®

Lieferant: Avantor
Gefahr

9017.2500EA 347 EUR
9017.2500 9017.1000 9017.5000 9017-413
Acetonitril ≥99,9%, BAKER ANALYZED® HPLC, Ultragradient für die HPLC, UHPLC, für die Spektrophotometrie, J.T.Baker®
Acetonitril
Formel: H₃CCN
Siedepunkt: 81,6 °C (1013 hPa)
Schmelzpunkt: –45,7 °C
Dichte: 0,782 g/cm³ (20 °C)
Flammpunkt: 2 °C (geschlossener Tiegel)
Lagertemperatur: Raumtemperatur
MDL: MFCD00001878
CAS-Nummer: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3,II
REACH: 01-2119471307-38
Merck Index: 13,00071
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cef8be50fcca82050c5bda827ad9c5f57f4c5e27&vid=b80b5c39e654fcf6904b18b58e22a816e8e138b7&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Meets Chromatography Reagent Specifications for testing Ph. Eur. monographs
Assay (by GC) min. 99.9 %
Appearance passes test
Distilling Range 80-82°C min. 95%
Identity (by IR) passes test
Transmittance 240 nm - 420 nm min. 98 %
Water (H2O) max. 0.01 %
Fluorescence Trace Impurities (as quinine base), ppb:
Measured at Emission Maximum for Solvent Impurities max. 1.0
Measured at 450 nm max. 0.3
Gradient Elution Test (PAH Suitability test) Ultraviolet Absorbance (a.u.)
at 254 nm max. 0.0005
Fluorescence (in ppb, measured as Benzo(a)Pyrene at EX/Em=280nm/Total Emission max. 0.5
at 210 nm max. 0.002
Limits of Impurities:
Residue after Evaporation max. 1 ppm
Titrable Acid (meq/g) max. 0.0008
Titrable Base (meq/g) max. 0.0006
Ultraviolet Absorbance (1.00-cm path vs water):
at 200 nm max. 0.05
at 220 nm max. 0.01
at 254-400 nm max. 0.01
UV Cut-off, nm max. 190


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR