Wasser Ph. Eur., USP, JP, gereinigt

Lieferant: VWR Chemicals

90200.9010EA 53.4 EUR
90200.9010 90200.9025 90200.9200 90200.0999 90200.5000
Wasser Ph. Eur., USP, JP, gereinigt
Wasser

  • Produced by reverse osmosis and capacitive deionization (CDI)


Bitte beachten Not for use as Active Pharmaceutical Ingredient (API)

Formel: H₂O
Siedepunkt: 100 °C (1013 hPa)
Schmelzpunkt: 0 °C
Dichte: 1,00 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011332
CAS-Nummer: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Bestellen

Spezifikation Testergebnisse

Appearance Clear colourless liquid
Heavy metals (as Pb) Max. 0.1 ppm
Total organic carbon (TOC) (on line) Max. 50 ppb
Yeasts and moulds (total count) Max. 100 CFU/ml
Bacterial endotoxins (IU/ml) Max. 0.25
Residual solvents (CPMP/ICH/283/95) Unlikely by manuf.process
Residues of metal catalysts or reagents Unlikely by manuf.process
As (Arsenic) Max. 0.1 ppm
Cd (Cadmium) Max. 0.1 ppm
Co (Cobalt) Max. 0.1 ppm
Hg (Mercury) Max. 0.1 ppm
Ni (Nickel) Max. 0.1 ppm
Pb (Lead) Max. 0.1 ppm
V (Vanadium) Max. 0.1 ppm
Conforms to Ph.Eur./USP/JP Passes test
Conforms to Ph.Eur. Monograph 0008 Passes test
Ph.Eur. specifications:
Total organic carbon (TOC) [Ph.Eur.] Max. 0.5 mg/l
Conductivity (20°C) (off line) [Ph.Eur.] ≤ 1.1 µS/cm
Elemental impurities (by conductivity) Passes test
Conductivity (20°C) (on line) [Ph.Eur.] Max. 1.1 μS/cm
Acidity or alkalinity Passes test Ph.Eur.
Oxidisable substances Passes test Ph.Eur.
Cl (Chloride) Passes test Ph.Eur.
SO4 (Sulphate) Passes test Ph.Eur.
NH4 (Ammonium) [Ph.Eur.] Max. 0.2 ppm
Calcium and magnesium Passes test Ph.Eur.
Residue on evaporation [Ph.Eur.] Max. 0.001 %
Microbial contamination Passes test Ph.Eur.
TAMC [Ph.Eur.] Max. 100 CFU/ml
USP specifications:
Total organic carbon [USP] Max. 0.50 mg/l
Water conductivity [USP] Max. 1.1 μS/cm
JP specifications:
Total organic carbon [JP] Max. 0.50 mg/l
Conductivity (25°C) [JP] Max. 2.1 μS/cm
KMnO4 reducing substances Passes test JP
Microbial limit Passes test JP
TAMC [JP] Max. 100 CFU/ml
Additional information
Bacterial endotoxins (Max. 0,25 IU/ml), tested on finished product Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR