Aceton 99,5-100,5% (durch Trockensubstanz) NF, Multikompendial, J.T.Baker®

Lieferant: Avantor
Gefahr

Synonyme: Propanon, DMK, 2-Propanon, Dimethylketon

9036-01EA 53.63 EUR
9036-01 9036-07 9036-03 9036-09
Aceton 99,5-100,5% (durch Trockensubstanz) NF, Multikompendial, J.T.Baker®
Aceton
Formel: CH₃COCH₃
Siedepunkt: 56,2 °C (1013 hPa)
Schmelzpunkt: –95,4 °C
Dichte: 0,792 g/cm³ (20 °C)
Flammpunkt: <–20 °C (geschlossener Tiegel)
Lagertemperatur: Raumtemperatur
MDL: MFCD00008765
CAS-Nummer: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3,II
REACH: 01-2119471330-49
Merck Index: 13,00067
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Meets E.P. Chemical Specifications
Meets B.P. Chemical Specifications
Meets N.F. Requirements
GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical Passes Test
NF - Identification A Passes Test
NF - Identification B Passes Test
NF - Specific Gravity at 25°/25°C ≤0.789
NF - Nonvolatile Residue ≤0.004 %
NF - Readily Oxidizable Substances Passes Test
NF - Water (H₂O) ≤0.5 %
NF - Assay (as (CH₃)₂CO) (calculated on anhydrous basis) 99.5 - 100.5 %
NF - Benzene ≤2 ppm
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Identification C Passes Test
EP/BP - Appearance of Solution Passes Test
EP/BP - Acidity or Alkalinity Passes Test
EP/BP - Density (g/mL) at 20°C 0.790 - 0.793
EP/BP - Related Substances - Impurity A ≤0.05 %
EP/BP - Related Substances - Impurity B ≤0.05 %
EP/BP - Related Substances - Impurity C ≤2 ppm
EP/BP- Other Impurities ≤0.05 %
EP/BP - Matter Insoluble in H₂O Passes Test
EP/BP - Reducing Substances Passes Test
EP/BP - Residue after Evaporation ≤50 ppm
EP/BP - Water (H₂O), g/L ≤3 g/l
Class 1, 2 and 3 residual solvents are likely to be present. Benzene (Class 1) is reported on each lot and is below the USP/NF limit concentration. Methanol and Toluene (Class 2) are both below their
Option 1 limits. All Class 3 solvents are <0.5%.
Elemental Impurities (USP (232), EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR