Benzylalkohol 98,0-100,5% NF, Multikompendial niedriger Peroxid-Gehalt, J.T.Baker®

Lieferant: Avantor
Achtung

Synonyme: Phenylcarbinol, (Hydroxymethyl)benzol, Phenylmethanol

9039-12EA 514.74 EUR
9039-12 9039-10 9039-14
Benzylalkohol 98,0-100,5% NF, Multikompendial niedriger Peroxid-Gehalt, J.T.Baker®
Benzylalkohol

Suitable for use in the manufacture of parenteral dosage forms.

Formel: C₆H₅CH₂OH
Siedepunkt: 205 °C (1013 hPa)
Schmelzpunkt: –15,3 °C
Dichte: 1,05 g/cm³ (20 °C)
Flammpunkt: 101 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004599
CAS-Nummer: 100-51-6
EINECS: 202-859-9
Merck Index: 13,01126
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0c85c958ac64f8b86c5836e49f2c4354b22886d&vid=633ad635461465914e782d5a22480c9ff8f81bf7&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

GMP Manufactured Product
Meets J.P. Chemical Specifications
Meets N.F. Requirements
Meets B.P. Chemical Specifications
Meets E.P. Chemical Specifications
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
NF - Assay 98.0 - 100.5 %
NF - Clarity of Solution Passes Test
NF - Color of Solution Passes Test
NF - Identification Passes Test
NF - Peroxide Value ≤5
NF - Related Substances - Benzaldehyde ≤0.05 %
NF - Related Substances - Cyclohexylmethanol ≤0.10 %
NF - Related Substances - Total Other Peaks ≤0.02 %
NF - Related Substances - Total other peaks >RT of benzyl alcohol ≤0.2 %
NF - Refractive Index, n²ºD 1.538 - 1.541
NF - Acidity Passes Test
NF - Residue on Evaporation ≤0.05 %
Solubility Passes Test
EP/BP - Assay 98.0 - 100.5 %
EP/BP - Identification Passes Test
EP/BP - Acidity Passes Test
EP/BP - Appearance of Solution Passes Test
EP/BP - Refractive Index, n²ºD 1.538 - 1.541
EP/BP - Peroxide Value ≤5
EP/BP - Residue on Evaporation ≤0.05 %
EP/BP - Related Substances - Benzaldehyde ≤0.05 %
EP/BP - Related Substances - Cyclohexylmethanol ≤0.10 %
EP/BP - Related Substances - Total Other Peaks ≤0.02 %
EP/BP - Related Substances - Total other peaks >RT of benzyl alcohol ≤0.2 %
JP - Assay 98.0 - 100.5 %
JP - Identification Passes Test
JP - Refractive Index, n²ºD 1.538 - 1.541
JP - Specific Gravity at 20°/20°C 1.043 - 1.049
JP - Clarity and Color of Solution Passes Test
JP - Acidity Passes Test
JP - Related Substances - Benzaldehyde ≤0.05 %
JP - Related Substances - Cyclohexylmethanol ≤0.10 %
JP - Related Substances - Total Other Peaks ≤0.02 %
JP - Related Substances - Total other peaks >RT of benzyl alcohol ≤0.2 %
JP - Peroxide Value ≤5
JP - Residue on Evaporation ≤0.05 %
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.
Suitable for use in the manufacture of parenteral dosage forms.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR