Methanol, J.T.Baker®

Lieferant: Avantor

9049.1000EA 59 EUR
9049.1000 9049.2500
Methanol, J.T.Baker®
Methanol
Formel: CH₃OH
MDL: MFCD00004595
CAS-Nummer: 67-56-1
UN: 1230
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Exceeds ACS Specifications
Assay (by GC) (corrected for water) min. 99.8 %
Appearance clear
Carbonyl Compounds (max. 0.001% each of Acetone, Formaldehyde and Acetaldehyde) passes test
Color (APHA) max. 10
Residue after Evaporation max. 0.001 %
Solubility in Water passes test
Substances Darkened by H2SO4 passes test
Substances Reducing KMnO4 passes test
Titrable Acid (meq/g) max. 0.0003
Titrable Base (meq/g) max. 0.0002
Water (H2O) max. 0.01 %
Trace Impurities (in ppm)
Aluminium (Al) max. 0.5
Boron (B) max. 0.02
Barium (Ba) max. 0.1
Calcium (Ca) max. 0.5
Cadmium (Cd) max. 0.05
Cobalt (Co) max. 0.02
Chromium (Cr) max. 0.02
Copper (Cu) max. 0.02
Iron (Fe) max. 0.1
Magnesium (Mg) max. 0.1
Manganese (Mn) max. 0.02
Nickel (Ni) max. 0.02
Lead (Pb) max. 0.1
Tin (Sn) max. 0.1
Zinc (Zn) max. 0.1


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR