Naphthol AS-D-chloracetat, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
Synonyme:
3-[[(2-Methylphenyl)amino]carbonyl]-2-naphthylchloracetat
M-Clarity™ Qualitätsstufe = MQ500
905-10CAP-S
905-10CAP-SEA
108
EUR
905-10CAP-S
Naphthol AS-D-chloracetat, Sigma-Aldrich®
Naphthol AS-D-chloracetat
Naphthol AS-D chloroacetate can be used as laboratory chemicals, manufacture of substances.
- Storage conditions: Tightly closed, dry
Formel:
C₂₀H₁₆ClNO₃ Molecular Weight: 353,8 g/mol |
CAS-Nummer:
35245-26-2 EINECS: 252-463-5 |
Spezifikation Testergebnisse
Appearance (colour) | White |
Appearance (form) | Crystalline Powder |
Performance | Conforms |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...