Tetrathiafulvalene-7,7,8,8-tetracyanoquinodimethane complex ≥97,0% (durch Elementaranalyse), Supelco®
Lieferant: Merck
Synonyme:
TTF - TCNQ Complex
M-Clarity™ Qualitätsstufe = MQ100
90548-1G-F
90548-1G-FEA
442
EUR
90548-1G-F
Tetrathiafulvalene-7,7,8,8-tetracyanoquinodimethane complex ≥97,0% (durch Elementaranalyse), Supelco®
Tetrathiafulvalene-7,7,8,8-tetracyanoquinodimethane complex
TTF-TCNQ is an organic metal-like charge transfer complex, which is mostly used as an electrode.[
TTF-TCNQ has been used in sensor fabrication during amperometric lactate biosensor development for qualitative analysis of lactate accumulation in ischemic myocardium patients.
Formel:
C₆H₄S₄ Molecular Weight: 408,53 g/mol |
MDL:
MFCD03791115 CAS-Nummer: 40210-84-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...