Supelco® TraceCERT® Organic Reference Standard, Methyl heptadecanoate
Lieferant: Merck
90606-100MG
90606-100MGEA 81.8 EUR
90606-100MG
Supelco® TraceCERT® Organic Reference Standard, Methyl heptadecanoate
Standards Organische Standards
Standards: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=37965a8044938c2cecdd71ad1248f04012fb63f1&vid=37965a8044938c2cecdd71ad1248f04012fb63f1&prid=web&clid=SAPDAM