Organic reference standard, Lithol Rubine BK, analytical standard, Supelco®
Lieferant: Merck
90689-25MG
90689-25MGEA
158
EUR
90689-25MG
Organic reference standard, Lithol Rubine BK, analytical standard, Supelco®
Standards
Organische Standards
Lithol rubine BK is a water soluble, synthetic red coloring dye, which may be used in drugs and cosmetics.
Verpackung: Bottomless glass bottle. Contents are inside inserted fused cone.