HPLC standards, LiChroTest®, Supelco®
Lieferant: Merck
1.19158.0001 1.19159.0001 1.19157.0001 1.19161.0001 1.19162.0001 1.19163.0001
909-0004EA 273 EUR
909-0004 909-0005 909-0002 909-0007 909-0008 909-0009
HPLC standards, LiChroTest®, Supelco®
Standards Gaschromatographie-Standards
Standard samples for HPLC System Qualification.
Zur Erleichterung dieser Qualifikationsarbeiten im HPLC-Labor wurden bei Avantor und Merck Millipore die LiChroTest®-Produkte entwickelt. Mit ihrer Hilfe können Sie die OQ und PQ routinemäßig, zeitsparend und nach einer standardisierten Methode durchführen.
The quality of analytical results is principally determined by the correct functioning of the analytical instruments employed. For this reason, the various quality assurance systems as well as the FDA require analytical instruments to be subjected to periodic qualification. Therefore, before starting a series of analyses, you first should establish whether your HPLC system meets with your requirements. These 'Operational Qualification' (OQ) and 'Performance Qualification' (PQ) steps involve tests of the different modules on their specifications and a check on the entire system using a real application relevant to the laboratory-specific requirements.