Schaeffer and Fulton Spore Stain Solution A, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Gefahr

Synonyme: Malachite Green Solution according to Schaeffer and Fulton

90903-100ML-F
90903-100ML-FEA 24.7 EUR
90903-100ML-F
Schaeffer and Fulton Spore Stain Solution A, Sigma-Aldrich®
Schaeffer and Fulton Spore Stain Solution A

In Schaeffer-Fulton`s method, a primary stain-malachite green is forced into the spore by steaming the bacterial emulsion. Malachite green is water soluble and has a low affinity for cellular material, so vegetative cells may be decolorized with water. Ve

MDL: MFCD06412418
CAS-Nummer: 2437-29-8
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR