Supelco® Organic Reference Standard, Conjugated (9E,11E)-Linoleic acid
Lieferant: Merck
90983-20MG
90983-20MGEA 322 EUR
90983-20MG
Supelco® Organic Reference Standard, Conjugated (9E,11E)-Linoleic acid
Standards Organische Standards
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bf3df9e66aea00cb3f4359b2e6abb9b466fde59d&vid=bf3df9e66aea00cb3f4359b2e6abb9b466fde59d&prid=web&clid=SAPDAM
Bestellen
TECHNISCHE DATEN
Synonym(s) (9E,11E)-9,11-Octadecadienoic acid; 9E,11E-CLA; Isolinoleic acid; Linoleic acid (9-trans, 11-trans); Mangold’s acid
M-Clarity Qualitätsstufe MQ100
Environmentally Preferable
CAS Nr. 544-71-8
MDL No. MFCD00133681
Molecular weight 280,45
Shipping conditions DICE
Lagerbedingungen –20 °C