Organic reference standard, Sucralose, analytical standard, Supelco®
Lieferant: Merck
Sucralose is a derivative of sucrose, which is generally used as a non-nutritive sweetener in foods and beverages.
A synthetic sweet tastant detectable by humans. Activates T1R2/T1R3 sweet taste receptors on enteroendocrine cells and elicits increased hormonal secretion of glucagon-like peptide-1 and glucose-dependent insulinotrophic peptide.
Verpackung: Bottomless glass bottle. Contents are inside inserted fused cone.