Phytochemical standard, Diosmetin, analytical standard, Supelco®
Lieferant: Merck
90985-10MG
90985-10MGEA 190 EUR
90985-10MG
Phytochemical standard, Diosmetin, analytical standard, Supelco®
Standards Organische Standards
It is a flavonoid obtained from Citrus limon. Act as an antioxidant. Inhibits carcinogen activation by inhibiting the CYP1A1 enzyme.
Verpackung: Bottomless glass bottle. Contents are inside inserted fused cone.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=62a5286b06b57c4aa50396a723850aa9099d9278&vid=62a5286b06b57c4aa50396a723850aa9099d9278&prid=web&clid=SAPDAM