Phytochemical standard, Tangeretin, analytical standard, Supelco®
Lieferant: Merck
91004-10MG
91004-10MGEA 242 EUR
91004-10MG
Phytochemical standard, Tangeretin, analytical standard, Supelco®
Standards Organische Standards
Tangeretin is a flavonoid found in the peel of citrus fruits where it most likely provides natural resistance to fungi. 
Tangeretin has been shown to counteract tumor promoter-induced inhibition of intercellular communication and to inhibit cell proliferation in several cancer lines.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1143ff52e88a17d0b3bbec55cafa1c2b7c285f7d&vid=1143ff52e88a17d0b3bbec55cafa1c2b7c285f7d&prid=web&clid=SAPDAM