Supelco® TraceCERT® Organic Reference Standard, Diisopropyl phthalate
Lieferant: Merck
91009-50MG
91009-50MGEA 85 EUR
91009-50MG
Supelco® TraceCERT® Organic Reference Standard, Diisopropyl phthalate
Standards Organische Standards
  • GHS signal: GHS08, GHS07
Standards: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Bitte beachten P201 - P302 + P352 - P305 + P351 + P338 - P308 + P313
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8cc6ad5c7aecf37849f1baab7931031ac0310598&vid=8cc6ad5c7aecf37849f1baab7931031ac0310598&prid=web&clid=SAPDAM