Naphthol AS-D-chloracetat-Lösung, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
Synonyme:
3-[[(2-Methylphenyl)amino]carbonyl]-2-naphthylchloracetat
911-10ML
911-10MLEA
177
EUR
911-10ML
Naphthol AS-D-chloracetat-Lösung, Sigma-Aldrich®
Naphthol AS-D-chloracetat
For use in Sigma Procedure 91C for the demonstration of specific esterase in blood, bone marrow films, tissue touch preparations, cytocentrifuge preparations and paraffin tissue sections.
Formel:
C₂₀H₁₆ClNO₃ Molecular Weight: 353,8 g/mol Lagertemperatur: Kühlschrank |
CAS-Nummer:
35245-26-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...