Anilin 99,0, BAKER ANALYZED®, J.T.Baker®

Lieferant: Avantor
Gefahr

Synonyme: Aminobenzol, Phenylamin

9110-01EA 204.43 EUR
9110-01
Anilin 99,0, BAKER ANALYZED®, J.T.Baker®
Anilin

Lot analysis on label.

Formel: C₆H₅NH₂
Molecular Weight: 93.13 g/mol
Siedepunkt: 184
Schmelzpunkt: –6.2 °C
Dichte: 1.019 g/cm³ (20 °C)
Flammpunkt: 76
Lagertemperatur: Raumtemperatur
MDL: MFCD00007629
CAS-Nummer: 62-53-3
EINECS: 200-539-3
UN: 1547
ADR: 6.1 II
Merck Index: 13,00661
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3fc5b5071f5a578057c8a7c1c4855ca964c4d840&vid=4f9c118a47c84e5b067d971f00a4eee6dc846d80&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

For Laboratory,Research,or Manufacturing Use = 99.0 %
NOTE: This product darkens to a reddish-brown colo r on storage. = 250
Assay (C₆H₅NH₂) (by GC, corrected for water) ≥99.0 %
Color (APHA) (at time of packaging) ≤250
Residue after Ignition ≤0.005 %
Chlorobenzene (C₆H₅Cl) ≤0.01 %
Hydrocarbons Passes Test
Nitrobenzene (C₆H₅NO₂) Passes Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR