VWR® PESTINORM®, GC Multiresidue Pesticide Standard (Mix 6) - 18 components, CRM
Lieferant: VWR Chemicals
A standard mixture, typically used for food testing, containing the following components: 100 ug/ml each of Acrinathrin [CAS:101007-06-1] ; Anthraquinone [CAS:84-65-1] ; cis-Bifenthrin [CAS:82657-04-3] ; Bioallethrin [CAS:584-79-2] ; Cyfluthrin [CAS:68359-37-5] ; lambda-Cyhalothrin [CAS:91465-08-6] ; Cypermethrin [CAS:52315-07-8] ; Deltamethrin [CAS:52918-63-5] ; Fenvalerate [CAS:51630-58-1] ; Flucythrinate [CAS:70124-77-5] ; tau-Fluvalinate [CAS:102851-06-9] ; cis-Permethrine [CAS:61949-76-6] ; trans-Permethrine [CAS:61949-77-7] ; Phenothrin [CAS:26002-80-2] ; Resmethrin [CAS:10453-86-8] ; Tefluthrin [CAS:79538-32-2] ; Tetramethrin [CAS:7696-12-0] ; Transfluthrin [CAS:118712-89-3] in Toluene
All mixes are supplied with detailed certificate of analysis, chromatogram and 100% shelf-life. The CoAs provide full traceability of preparation, including the dissolution of the raw materials (with their purity), the preparation of the intermediate solutions and the preparation of the final solutions.
Standards: Certified Reference Material (CRM) acc. ISO 17025 / ISO 17034
Bitte beachten Storage temperature: Freezer (–20 °C)