VWR®, Alternate Metals II, 4 components: Ca 500 mg/l, Na 500 mg/l, K 100 mg/l, Mg 100 mg/l in 2% HNO₃
Lieferant: VWR Chemicals
91174.180EA
217
EUR
91174.180
91174.260
VWR®, Alternate Metals II, 4 components: Ca 500 mg/l, Na 500 mg/l, K 100 mg/l, Mg 100 mg/l in 2% HNO₃
Standards
Kationischer Standards
Mehrelement-Standards
Standards: ISO 17025/17034