Fast Red Violet LB Base solution, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Achtung

912-10ML
912-10MLEA 194 EUR
912-10ML
Fast Red Violet LB Base solution, Sigma-Aldrich®
Fast Red Violet LB Base solution

Used as a kit component in Procedure 91 for the demonstration of specific esterase activity in peripheral blood smears or paraffin sections.

Lagertemperatur: Kühlschrank
MDL: MFCD00007780
ADR: 8,III
IVDLogo

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR