Supelco® TraceCERT® Organic Reference Standard, Gallic acid
Lieferant: Merck
91215-100MG
91215-100MGEA 90.1 EUR
91215-100MG
Supelco® TraceCERT® Organic Reference Standard, Gallic acid
Standards Organische Standards
Standards: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Bitte beachten P261 - P264 - P271 - P280 - P302 + P352 - P305 + P351 + P338
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e76251fa3a3f94317236616ad650827b694d49bf&vid=e76251fa3a3f94317236616ad650827b694d49bf&prid=web&clid=SAPDAM