VWR®, GF AAS Multi element standard XVIII, 16 components in 5% HNO₃
Lieferant: VWR Chemicals
91265.180EA
289
EUR
91265.180
VWR®, GF AAS Multi element standard XVIII, 16 components in 5% HNO₃
Standards
Kationischer Standards
Mehrelement-Standards
16 components: Ag 10 mg/l, Al 100 mg/l, As 100 mg/l, Ba 50 mg/l, Be 5 mg/l, Cd 5 mg/l, Co 50 mg/l, Cr 20 mg/l, Cu 50 mg/l, Fe 20 mg/l, Mn 20 mg/l, Ni 50 mg/l, Pb 100 mg/l, Sb 100 mg/l, Se 100 mg/l, Tl 100 mg/l in 5% HNO₃
Standards: ISO 17025/17034