VWR®, ICP Calibration Standard - Quality Control (XVI), 21 Elements: 100 mg/l each of As, Be, Ca, Cd, Co, Cr, Cu, Fe, Li, Mg, Mn, Mo, Ni, Pb, Sb, Se, Sr, Ti, Tl, V, Zn in 2% HNO₃
Lieferant: VWR Chemicals
91266.180EA 385 EUR
91266.180
VWR®, ICP Calibration Standard - Quality Control (XVI), 21 Elements: 100 mg/l each of As, Be, Ca, Cd, Co, Cr, Cu, Fe, Li, Mg, Mn, Mo, Ni, Pb, Sb, Se, Sr, Ti, Tl, V, Zn in 2% HNO₃
Standards Kationischer Standards Mehrelement-Standards
Standards: ISO 17025/17034