Ammoniumdihydrogenphosphat 20 g/L in Salpetersäure 1%, ARISTAR® Matrixmodifizierlösung, für die Graphitofen-AAS

Lieferant: VWR Chemicals
Achtung

Synonyme: Ammonium phosphate, monobasic, Ammonium Phosphate Monobasic, Ammonium dihydrogenphosphate

91276.154EA 91.4 EUR
91276.154
Ammoniumdihydrogenphosphat 20 g/L in Salpetersäure 1%, ARISTAR® Matrixmodifizierlösung, für die Graphitofen-AAS
Ammoniumdihydrogenphosphat

The VWR® Matrix modifier solutions have especially been developed for use in GF-AAS to eliminate matrix effects. They can be used either diluted or as mixtures.


  • Guaranteed accuracy: ±1%
  • COA incl. up to 70 measured trace impurities in ppm range


VWR® Matrix modifier solutions are produced with high purity metals or salts (≥99,999%) in sub-boiling distilled acids and are supplied with a detailed batch specific certificate of analysis.

Formel: NH₄H₂PO₄
Molecular Weight: 115,03 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00003396
CAS-Nummer: 7722-76-1
UN: 1760
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR