Palladium matrix modifier 5 g/L in Salpetersäure 1% [99,999% Pd(NO₃)₂], ARISTAR® für die Graphitofen-AAS
Lieferant: VWR Chemicals
|
Achtung
|
The VWR® Matrix modifier solutions have especially been developed for use in GF-AAS to eliminate matrix effects. They can be used either diluted or as mixtures.
- Guaranteed accuracy: ±1%
- COA incl. up to 70 measured trace impurities in ppm range
VWR® Matrix modifier solutions are produced with high purity metals or salts (≥99,999%) in sub-boiling distilled acids and are supplied with a detailed batch specific certificate of analysis.
Lagertemperatur:
Raumtemperatur |
UN:
1760 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...