Multi cation standard 1 for IC, TraceCERT®, Supelco®
Lieferant: Merck
91286-100ML
91286-100MLEA 220 EUR
91286-100ML
Multi cation standard 1 for IC, TraceCERT®, Supelco®
Standards Kationischer Standards Mehrelement-Standards
Ca, 1000 mg/l
K, 200 mg/l
Li, 50 mg/l
Mg, 200 mg/l
Na, 200 mg/l
NH4, 400 mg/l
Standards: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034