VWR®, Mixed Anions Standard, 7 components in H₂O
Lieferant: VWR Chemicals
91296.180EA
190
EUR
91296.180
VWR®, Mixed Anions Standard, 7 components in H₂O
Standards
Anionische Standards
Anionische Multi-Element-Standards
7 components: Fluorides (F-) 2ppm, Phosphates (PO43-) 10ppm, Nitrites (NO2-) 5ppm, Nitrates (NO3-) 10ppm, Bromides (Br-) 10ppm, Chlorides (Cl-) 5ppm, Sulphates (SO42-) 10ppm in H₂O
Standards: ISO 17025/17034