Organic reference standard, (2RS)-2-(4-Hydroxyphenyl)propionic acid, ≥95,0% (qNMR), Supelco®
Lieferant: Merck
91378-10MG
91378-10MGEA
39.5
EUR
91378-10MG
Organic reference standard, (2RS)-2-(4-Hydroxyphenyl)propionic acid, ≥95,0% (qNMR), Supelco®
Standards
Organische Standards
Biochemical and physiol actions are 2-(4-Hydroxyphenyl)propionic acid is a polyphenol metabolite detected in biological fluids. This compound belongs to the class of organic compounds known as phenylpropanoic acids. These are compounds with a structure containing a benzene ring conjugated to a propanoic acid.