tri-Natriumcitrat-Lösung, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

915-50ML
915-50MLEA 105 EUR
915-50ML
tri-Natriumcitrat-Lösung, Sigma-Aldrich®
tri-Natriumcitrat

Used as a kit component for the 91A and 91C procedures for Esterase.

Formel: C₆H₅Na₃O₇
Lagertemperatur: Kühlschrank
MDL: MFCD00012462
CAS-Nummer: 68-04-2
IVDLogo

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR