1-Naphthylacetat-Lösung, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
|
Gefahr
|
Synonyme:
Essigsäure(1-naphthyl)ester, Essigsäure(α-naphthyl)ester, α-Naphthylacetat
916-10ML
916-10MLEA
182
EUR
916-10ML
1-Naphthylacetat-Lösung, Sigma-Aldrich®
1-Naphthylacetat
Reagent used in the 91A procedure for the demonstration of non-specific esterase activity in leukocytes.
Formel:
C₁₂H₁₀O₂ Molecular Weight: 186,21 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00003922 CAS-Nummer: 830-81-9 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...