Tridodecylmethylammonium chloride, Selectophore™ Ionophor, Supelco®

Lieferant: Merck
Achtung

Synonyme: Methyltridodecylammonium chloride

M-Clarity™ Qualitätsstufe = MQ100

91661-1G
91661-1GEA 258 EUR
91661-1G
Tridodecylmethylammonium chloride, Selectophore™ Ionophor, Supelco®
Tridodecylmethylammonium chloride

Electroactive component in chloride-selective solvent polymeric membrane electrode


Verpackung: Bottomless glass bottle. Contents are inside inserted fused cone.

Formel: [CH₃(CH₂)₁₁]₃N(Cl)CH₃
Molecular Weight: 572.47 g/mol
MDL: MFCD00043303
CAS-Nummer: 7173-54-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da964a7971e4a3b88cd8978149750ec6347b270a&vid=000a3d87683bc47b2cdcbc7784124bf631391312&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR