Tridodecylmethylammonium nitrate ≥99,0%, Selectophore™ Ionophor, Supelco®
Lieferant: Merck
|
Achtung
|
Synonyme:
Methyltridodecylammonium nitrate
M-Clarity™ Qualitätsstufe = MQ100
91664-1G
91664-1GEA
540
EUR
91664-1G
Tridodecylmethylammonium nitrate ≥99,0%, Selectophore™ Ionophor, Supelco®
Tridodecylmethylammonium nitrate
Tridodecylmethylammonium nitrate (TDMAN) is a nitrate ionophore which has good selectivity for nitrate more than chloride.
TDMAN has been used in preparing liquid ion-selective electrode membrane during coulometric analysis of nitrate from fresh water samples.
Verpackung: Bottomless glass bottle. Contents are inside inserted fused cone.
Formel:
[CH₃(CH₂)₁₁]₃N(NO₃)CH₃ Molecular Weight: 599,03 g/mol |
MDL:
MFCD00145447 CAS-Nummer: 13533-59-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...