Glove bags, small
Lieferant: Thermo Fisher Scientific
91717.LK 91717.LL
91717.LKEA 126 EUR
91717.LK 91717.LL
Glove bags, small
Handschuhboxen und -beutel
These polyethylene glove bags are ideal for laboratory use and field sampling.
Suitable for handling air and moisture sensitive, high purity chemicals and materials. Soluble in xylene.
Lieferumfang: Supplied with closure, tubing coupler, and cotton gloves.