Phytochemical standard, Cynarin, analytical standard, Supelco®
Lieferant: Merck
91801-5MG
91801-5MGEA
314
EUR
91801-5MG
Phytochemical standard, Cynarin, analytical standard, Supelco®
Standards
Organische Standards
Phenolic compound found in artichoke. Antioxidant and choleretic properties. Potential immunosuppressive agent.
Verpackung: Bottomless glass bottle. Contents are inside inserted fused cone.