Phytochemical standard, Cynarin, analytical standard, Supelco®
Lieferant: Merck
91801-5MG
91801-5MGEA 314 EUR
91801-5MG
Phytochemical standard, Cynarin, analytical standard, Supelco®
Standards Organische Standards
Phenolic compound found in artichoke. Antioxidant and choleretic properties. Potential immunosuppressive agent.
Verpackung: Bottomless glass bottle. Contents are inside inserted fused cone.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f1a85720cae887004ba2ea6bccc7c167f25fa32d&vid=f1a85720cae887004ba2ea6bccc7c167f25fa32d&prid=web&clid=SAPDAM