Chloroform ≥99.8% (by GC, corrected for water content) stabilised, BAKER ANALYZED® ACS, J.T.Baker®

Lieferant: Avantor
Gefahr

9186.1000EA 133.63 EUR
9186.1000 9186.2500 9186.9200
Chloroform ≥99.8% (by GC, corrected for water content) stabilised, BAKER ANALYZED® ACS, J.T.Baker®
Chloroform
Formel: CHCl₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00000826
CAS-Nummer: 67-66-3
EINECS: 200-663-8
UN: 1888
REACH: 05-2114614906-45

Bestellen

Spezifikation Testergebnisse

Assay (by GC) min. 99.8 %
Acetone and Aldehyde (as(CH3)2CO) max. 0.005 %
Acid and Chloride passes test
Color (APHA) ≤ 10
Free Chlorine passes test
Preservative (C2H5OH) about 0.75%
Residue after Evaporation max. 0.001 %
Substances Darkened by H2SO4 passes test
Suitability for Use with Dithizone Test passes test
Trace Impurities (in ppm)
Aluminium (Al) max. 0.5
Barium (Ba) max. 0.1
Boron (B) max. 0.02
Cadmium (Cd) max. 0.05
Calcium (Ca) max. 0.5
Chromium (Cr) max. 0.02
Cobalt (Co) max. 0.02
Copper (Cu) max. 0.02
Iron (Fe) max. 0.1
Lead (Pb) max. 0.05
Magnesium (Mg) max. 0.1
Manganese (Mn) max. 0.02
Nickel (Ni) max. 0.02
Tin (Sn) max. 0.1
Zinc (Zn) max. 0.1


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR